
Dichloromethane-[D₂], Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1665-00-5
:
Formula:CD₂Cl₂
MDL Number:MFCD00000882
:86.92 g/mol
Boiling Pt:40 °C (1013 hPa)
Melting Pt:–97 °C
Density:1.36 g/cm³ (20 °C)
EINECS:216-776-0
ADR:6.1,III
:M-Clarity™ quality level = MQ100
UN:1593
:GHS08
Storage Temperature:Ambient
Frequently Bought Together