Supelco® PESTANAL® Organic Reference Standard, Cycloheximide
Catalog # 46401-100MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:66-81-9
- Description:Cycloheximide
- Seq:2440
- Storage conditions:–20 °C
- Hazard class description:H300 - H341 - H360D - H411
- Shipping conditions:WICE
- MDL No.:MFCD00082346
- M-Clarity quality level:MQ100
- Synonym(s):3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide; Actidione; Naramycin A
- Molecular weight:281,35
Specifications
About this item
- GHS signal: GHS06, GHS08, GHS09
Caution: P202 - P264 - P270 - P273 - P280 - P301 + P310