Order Entry
Slovakia
ContactUsLinkComponent
Hexane (mixture of isomers) ≥95%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Irritant
Health
Flammable
Environment
Hexane (mixture of isomers) ≥95%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 650552-1L
Supplier:  HONEYWELL
CAS Number:  
undefined
Irritant
Health
Flammable
Environment
Hexane (mixture of isomers) ≥95%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 650552-1L
Supplier:  HONEYWELL
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Vapor Pressure:
    256 mmHg ( 37.7 °C)
  • Assay (GC):
    Min. 95.0 %
  • Non-volatile matter:
    Max. 0.0001 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Refractive index (n 20/D):
    1.373 - 1.377
  • Absorbance at 400 nm:
    Max. 0.005
  • Absorbance 280 nm:
    Max. 0.005
  • Absorbance at 250 nm:
    Max. 0.01
  • Absorbance at 220 nm:
    Max. 0.05
  • Absorbance at 210 nm:
    Max. 0.20
  • Absorbance at 200 nm:
    Max. 0.50
  • Absorbance at 195 nm:
    Max. 1.0
  • Fluorescence (chinin) at 365 nm:
    Max. 1 ppb
  • IR spectrum:
    complying
  • GC (as heptachlor epoxide):
    Max. 10 ng/l
  • Autoignition Temperature:
    453 °F
  • Refractive Index:
    n20/D 1.375 (lit.)

Specification Test Results

About this item

 :
 :86.18 g/mol
Merck Index:13,04712
Boiling Pt:66…69 °C(1.013 hPa)
Melting Pt:–95 °C
Density:0.660…0.668 g/cm³ (20 °C)
Flash Pt:7.4 %(V)
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520