Neck tube, FR19
Supplier: Portwest
Certificates
About this item
This anti-static neck tube is flame resistant so can be worn when dealing with flame or heat. Made from Modaflame knit (60% Modacrylic, 39% cotton and 1% carbon fibre).
- Ideal for protection in multiple work places
- Extremely comfortable
- Provides protection for the entire neck area
Weight: 200 g/m²
Specifications
- Catalog No:
- PRTWFR19NAR
- PRTWFR19BKR
- Model:
- FR19
- FR19
- Garment application:
- Fire protection, Industrial
- Fire protection, Industrial
- Risk area:
- Heat, Flame-retardant, Anti-static
- Heat, Flame-retardant, Anti-static
- Weight:
- 200 g/m²
- 200 g/m²