Order Entry
Slovakia
ContactUsLinkComponent
Ethyl acetate 99.8-100.5% BP, Ph. Eur., pharma grade
Flammable
Irritant
Ethyl acetate 99.8-100.5% BP, Ph. Eur., pharma grade
Catalog # APPC191318.0616
Supplier:  PanReac AppliChem
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
Ethyl acetate 99.8-100.5% BP, Ph. Eur., pharma grade
Catalog # APPC191318.0616
Supplier:  PanReac AppliChem
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pk:
    25 L
  • Seq:
    5327
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (C₄H₈O₂):
    99.8 - 100.5%
  • Identity according to Pharmacopoeias:
    Passes Test
  • Refractive Index n20/D:
    1.370 - 1.373
  • Appearance of Solution:
    Passes Test
  • Acidity:
    Passes Test
  • Insoluble Matter in H₂O:
    Passes Test
  • Non-Volatile Matter:
    Max. 0.003%
  • Darkened Substances by H₂SO₄:
    Passes Test
  • Related Substances:
    Passes Test
  • Residual Solvents (Ph. Eur.):
    Passes Test
  • Water (H₂O):
    Max. 0.05%
  • Cadmium (Cd) - ICH Q3D:
    Max. 0.5 ppm
  • Lead (Pb) - ICH Q3D:
    Max. 0.5 ppm
  • Arsenic (As) - ICH Q3D:
    Max. 1.5 ppm
  • Mercury (Hg) - ICH Q3D:
    Max. 1.5 ppm
  • Cobalt (Co) - ICH Q3D:
    Max. 5 ppm
  • Vanadium (V) - ICH Q3D:
    Max. 10 ppm
  • Nickel (Ni) - ICH Q3D:
    Max. 20 ppm
  • Thallium (Tl) - ICH Q3D:
    Max. 0.8 ppm
  • Gold (Au) - ICH Q3D:
    Max. 10 ppm
  • Palladium (Pd) - ICH Q3D:
    Max. 10 ppm
  • Iridium (Ir) - ICH Q3D:
    Max. 10 ppm
  • Osmium (Os) - ICH Q3D:
    Max. 10 ppm
  • Rhodium (Rh) - ICH Q3D:
    Max. 10 ppm
  • Ruthenium (Ru) - ICH Q3D:
    Max. 10 ppm
  • Selenium (Se) - ICH Q3D:
    Max. 15 ppm
  • Silver (Ag) - ICH Q3D:
    Max. 15 ppm
  • Platinum (Pt) - ICH Q3D:
    Max. 10 ppm
  • Lithium (Li) - ICH Q3D:
    Max. 55 ppm
  • Antimony (Sb) - ICH Q3D:
    Max. 120 ppm
  • Barium (Ba) - ICH Q3D:
    Max. 140 ppm
  • Molybdenum (Mo) - ICH Q3D:
    Max. 25 ppm
  • Copper (Cu) - ICH Q3D:
    Max. 250 ppm
  • Tin (Sn) - ICH Q3D:
    Max. 600 ppm
  • Chromium (Cr) - ICH Q3D:
    Max. 25 ppm

Specification Test Results

About this item

 :
 :88.11 g/mol
Boiling Pt:77 °C
Melting Pt:–83 °C
Density:0.902 g/cm³
Flash Pt:–4 °C
EINECS:205-500-4
UN:1173
ADR:3,II
 :GHS02
 :GHS07
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171