E-64-C
Catalog # ENZOBMLPI1060005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:03
- Protein/peptide name:E-64-C
- Purity:98%
Specifications
About this item
Calpain and cathepsin inhibitor
Cell-impermeable epoxysuccinyl peptide inhibitor of calpain and other cysteine proteases. Inhibits calpain and cathepsins B, H, and L.