PKCzeta (113-125) (myristoylated)
Catalog # ENZOBMLP2190500
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:0,5 mg
- Seq:02
- Protein/peptide name:PKCzeta (113-125) (myristoylated)
- Purity:>97% (HPLC)
- Formulation:Lyophilised
Specifications
About this item
PKC inhibitor
Pseudosubstrate sequence from human PKCzeta (113-125) completely conserved in rabbit, rat, mouse and Xenopus. Sequence is also identical in human PKCι; (114-126) and mouse PKClambda. The N-terminus is myristoylated to allow cell permeability. Peptide inhibitor of atypical PKC's (calcium/phorbol ester/diacyglycerol insensitive). Useful in conjunction with the Myristoylated PKC (20-28), Cell-permeable PKC Inhibitor (BML-P205).