Human recombinant C-reactive protein
ENZOENZPRT1640100
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Protein synonyms:CRP|PTX1
- Protein/peptide name:C-reactive protein
- Purity:>95% (SDS-PAGE)
- Molecular weight:~24 kDa
- Formulation:Lyophilized from 0,22 µm filtered solution in PBS. Generally 5 - 8% mannitol or trehalose is added as a protectant before lyophilization.
Produced in HEK 293 cells.