Mouse recombinant IL-1beta
Catalog # ENZOENZPRT1360010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Seq:02
- Protein synonyms:Catabolin|Interleukin-1β
- Protein/peptide name:IL-1beta
- Purity:>98% (SDS-PAGE gel & HPLC)
- Molecular weight:~17.5 kDa
- Formulation:Lyophilized from 10 mm sodium citrate, pH 4,0
Specifications
About this item
Produced in E.coli.