Human recombinant PDE5A
ENZOENZPRT1260010
:
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Protein synonyms:cGMP-specific 3'|Phosphodiesterase 5A|CGB-PDE|5'-cyclic phosphodiesterase
- Protein/peptide name:PDE5A
- Purity:>95%
- Formulation:Liquid. In 50 mm triethanolamine, pH 7,4, containing 1 mm DTT, 100 mm NaCl, 20% glycerol
Produced in Sf9 insect cells. Recombinant human PDE5A (phosphodiesterase 5A) is fused at the C-terminus to a His-tag.