Recombinant dengue NS1 ST3
Catalog # ENZOENZPRT1060100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Seq:02
- Protein synonyms:non-structural protein|viral|strain Sri Lanka D3/H/IMTSSA-SRI/2000/1266|DENV1-NS1
- Protein/peptide name:Dengue NS1 ST3
- Purity:>95% (SDS-PAGE)
- Formulation:Liquid, In DPBS pH 7,4
- Purification:Purified protein
Specifications
About this item
Dengue virus NS1 protein serotype 3 presented in its native folded state with all post-translational modifications to deliver optimal antigenicity.
Produced in HEK 293 cells. Dengue Viral NS1 Serotype 3 (DENV1-NS1) from strain Sri Lanka D3/H/IMTSSA-SRI/2000/1266.