Order Entry
Slovakia
ContactUsLinkComponent
Human native beta2-Microglobulin
Human native beta2-Microglobulin
Catalog # ENZOALX5224610100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human native beta2-Microglobulin
Catalog # ENZOALX5224610100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Native
  • Species:
    Human
  • Seq:
    02
  • Protein synonyms:
    MHC class I|B2M protein
  • Protein/peptide name:
    beta2-Microglobulin
  • Purity:
    >95% (SDS-PAGE)
  • Molecular weight:
    ~12kDa
  • Formulation:
    Liquid. In PBS, pH 7,2, containing 150 mM sodium chloride, 1 mM EDTA sodium salt and 0,05% sodium azide.

Specifications

About this item

High purity, native protein used as a target diagnostic marker for cancer or renal failure.

Purified native human β2-Microglobulin protein. Purified from urine of patients with chronic renal tubular proteinuria.