Human recombinant Gabarap, GST tag
Catalog # ENZOBMLUW11750500
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:500 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Tag sequence:GST Tag
- Protein/peptide name:Gabarap
- Purity:>90% (SDS-PAGE)
- Molecular weight:~42,1kDa
- Concentration:0,36 mg/ml
- Formulation:Liquid. In PBS.
Specifications
About this item
Produced in E. coli.