Native C. Trachomatis
Catalog # ENZOALX6301241000
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mL
- Protein/peptide type:Native
- Seq:02
- Protein/peptide name:C. Trachomatis
- Formulation:Liquid, PBS containing 0,05% Triton X-100
- Purification:Cell lysate.
Specifications
About this item
Cell lysate ideal for use as a broad positive control.
From Chlamydia trachomatis, serotype LGV2 strain 434. Inactivated by heat treatment and 0.05% Triton X-100.