Human recombinant ranGAP1 fragment (GST-tag) (from E. coli), GST Tag
ENZOBMLUW97550100
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:GST Tag
- Protein/peptide name:RanGAP1 fragment (GST-tag)
- Formulation:Liquid. In 50 mM TRIS, pH 7,5, containing 0,5 mM dithiothreitol and 150 mM sodium chloride.
Produced in E. coli.