Order Entry
Slovakia
ContactUsLinkComponent
 
Human recombinant DPP8
Catalog # ENZOBMLSE5270010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant DPP8
Catalog # ENZOBMLSE5270010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    DPP8

Specifications

About this item

Produced in insect cells. Active recombinant human DPP8 with an N-terminal purification tag.