Order Entry
Slovakia
ContactUsLinkComponent
Human recombinant PDE4D (catalytic domain) (from E. coli)
Human recombinant PDE4D (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE5230020
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant PDE4D (catalytic domain) (from E. coli)
Catalog # ENZOBMLSE5230020
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    PDE4D (catalytic domain)

Specifications

About this item

Produced in E. coli. Active catalytic domain of phosphodiesterase 4D.