Order Entry
Slovakia
ContactUsLinkComponent
Human recombinant PKCtheta
Human recombinant PKCtheta
Catalog # ENZOBMLSE5170005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant PKCtheta
Catalog # ENZOBMLSE5170005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    PKCtheta

Specifications

About this item

Produced in insect cells. Full length active enzyme fused at the N-terminus to a GST-tag.