Human recombinant Csk (GST-tag),GST Tag
ENZOBMLSE3800005
:
- Pk:5 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Tag sequence:GST Tag
- Protein/peptide name:Csk (GST-tag)
Produced in insect cells. Full length active enzyme.
Produced in insect cells. Full length active enzyme.