Human recombinant Tau-352 (0N3R variant) (from E. coli)
Catalog # ENZOBMLSE3220100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:Tau-352 (0N3R variant)
- Formulation:Lyophilised
Specifications
About this item
Produced in E. coli.