Order Entry
Slovakia
ContactUsLinkComponent
Human recombinant Calcineurin (from E. coli)
Human recombinant Calcineurin (from E. coli)
Catalog # ENZOBMLSE1635000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant Calcineurin (from E. coli)
Catalog # ENZOBMLSE1635000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 Vial
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    Calcineurin
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    0,09 mg/ml
  • Formulation:
    100 U/µl, in 1x KI-128 Assay Buffer with increased DTT concentration (50 mM TRIS, pH 7,5, containing 100 mM sodium chloride, 6 mM MgCl2, 5 mM DTT, 0,025% NP-40 and 0,5 mM CaCl2)

Specifications

About this item

Produced in E.coli. Human calcineurin Aalpha (MW 60kDa) and calcineurin B (MW 15kDa) heterodimer.

Both subunits are coexpressed in a construct with yeast myristoyl-CoA:protein N-myristoyltransferase. The resulting highly active calcineurin (protein phosphatase 2B) is N-myristoylated on the CaNB subunit, similar to the native protein.