Human recombinant LAR (from E. coli)
Katalogové číslo ENZOBMLSE1130200
Dodavatel: ENZO LIFE SCIENCES
Číslo CAS:
Specifikace
- Pk:1 Vial
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:LAR
- Purity:>90% (SDS-PAGE)
- Formulation:Liquid. In 50 mM TRIS-HCl, pH 7,0, containing 100 mM sodium chloride, 2 mM EDTA, 5mM dithiothreitol, 001% Brij 35, and 30% glycerol.
- Purification:Contains no detectable protease, DNase, RNase, or Ser/Thr phosphatase activity.
Specifikace
O tomto produktu
Produced in E. coli. Soluble catalytic LAR-D1 domain (350 aa).