PDKtide
Catalog # ENZOBMLP2500100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Seq:02
- Protein/peptide name:PDKtide
- Purity:95%
Specifications
About this item
Protein kinase substrate
This peptide, KTFCGTPEYLAPEVRREPRILSEEEQEMFRDFDYIADWC, is derived from AKT1 and PKN2/PRK2. It is a substrate for PDK1, CDK9/Cyclin T1 and JAK2.