Order Entry
Slovakia
ContactUsLinkComponent
Protein tyrosine phosphatase substrate
Protein tyrosine phosphatase substrate
  ENZOBMLP1530001
 :  
Protein tyrosine phosphatase substrate
  ENZOBMLP1530001
 :  
  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    Protein tyrosine phosphatase substrate
This peptide sequence, ENDpYINASL, is derived from a highly conserved region of the T-cell phosphatase, TCPTP, and has been shown to be an excellent general substrate for protein tyrosine phosphatases.