OmniMMP™ fluorogenic control
Catalog # ENZOBMLP1270001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:OmniMMP™ fluorogenic control
- Purity:98%
- Tested applications:Control
Specifications
About this item
Reference peptide for OmniMMPTM fluorogenic substrate (BML-P126) and TACE (ADAM17) fluorogenic substrate (BML-P132). Ex.: 328 nm, Em.: 393 nm.
The following Ex/Em have also been used: 320 to 340/400 to 420.