Order Entry
Slovakia
ContactUsLinkComponent
 
Human recombinant CD40L, soluble
  ENZOALX850064KI01
 :  
Human recombinant CD40L, soluble
  ENZOALX850064KI01
 :  

 

  • Pk:
    1 SET
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Reconstitution instructions:
    Reconstitute CD40L with 100 µl and each vial of enhancer with 50 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or carrier protein
  • Protein/peptide name:
    CD40L, soluble
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    CD40L: 0,1 mg/ml after reconstitution
    Enhancer: 1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

 

 

Can be used to activate human B cells and dendritic cells. Stimulates growth of human B cells. For stimulation of mouse cells via CD40 use MAb to CD40 (mouse) (FGK45) (Prod. No. ALX-805-046).

 : Contents: 10 µg of CD40L, Soluble (human) (recombinant) (Prod. No. ALX-522-015) and 2×50 µg of Enhancer for Ligands (Prod. No. ALX-804-034), which increases the biological activity of rhsCD40L at least 1000-fold.