Rat Recombinant Omentin
Catalog # ENZOALX522132C010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Rat
- Seq:02
- Reconstitution instructions:Reconstitute with 100 µl sterile water.
- Protein/peptide name:Omentin
- Purity:>95% (SDS-PAGE)
- Concentration:0,1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Specifications
About this item
Produced in HEK 293 cells. Rat omentin (aa 20-313) is fused at the N-terminus to a FLAG®-tag.