Order Entry
Slovakia
ContactUsLinkComponent
Rat recombinant IL33 (from E. coli)
Rat recombinant IL33 (from E. coli)
  ENZOALX522125C010
 :  
Rat recombinant IL33 (from E. coli)
  ENZOALX522125C010
 :  

 

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Seq:
    02
  • Reconstitution instructions:
    Reconstitute with 100 µl sterile distilled water. Further dilutions should be made with medium containing 5% fetal calf serum.
  • Protein/peptide name:
    IL33
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    0,1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

 

 

Produced in E. coli. Untagged rat IL-33 (aa 109-264).