Mouse recombinant LAG-3:Fc (from CHO cells)
Catalog # ENZOALX522099C050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Mouse
- Seq:02
- Biological activity:Induces maturation of mouse dendritic cells in vitro. Induces CD4 and CD8 antigen-specific T cells responses when coinjected s.c. in mice, together with the antigen.
- Reconstitution instructions:Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
- Protein/peptide name:LAG-3:Fc
- Purity:>95% (SDS-PAGE)
- Concentration:0,5 mg/ml
- Formulation:Liquid. In PBS.
Specifications
About this item
Produced in CHO cells. The sequence coding for the 4 extracellular Ig-like domains of mouse LAG-3 (D1-D4) is fused to the Fc portion of mouse IgG2a (hinge-CH2-CH3).