Order Entry
Slovakia
ContactUsLinkComponent
Mouse recombinant BAFF (soluble)
Mouse recombinant BAFF (soluble)
Catalog # ENZOALX522052C010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Mouse recombinant BAFF (soluble)
Catalog # ENZOALX522052C010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Seq:
    02
  • Biological activity:
    Mediates splenocyte survival.
  • Reconstitution instructions:
    Reconstitute with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or other carrier protein.
  • Protein/peptide name:
    BAFF (soluble)
  • Purity:
    >90% (SDS-PAGE)
  • Concentration:
    0,1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

Specifications

About this item

Produced in HEK 293 cells. The extracellular domain of mouse BAFF (aa 127-309) is fused at the N-terminus to a linker peptide (10 aa) and a FLAG®-tag.