Order Entry
Slovakia
ContactUsLinkComponent
Ac-VDVAD-pNA
Ac-VDVAD-pNA
Supplier:  ENZO LIFE SCIENCES
Certificates
Ac-VDVAD-pNA
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide name:
    Ac-VDVAD-pNA
  • Purity:
    >98% (HPLC)
  • Formulation:
    Lyophilized

Specifications

Frequently Bought Together