Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Poly(ADP-ribose), standard

Poly(ADP-ribose), standard

1 µG
Catalog # ENZOALX202043C001
Supplier:   Enzo Life Sciences
Poly(ADP-ribose), standard
Catalog # ENZOALX202043C001
Supplier:  Enzo Life Sciences
CAS Number:  
Poly(ADP-ribose), standard

1 µG

CAS Number:  
Price
542,54 €
Log in for availability
1 * 1 µG
Product Details & Documents

Released from automodified PARP-1 by base treatment and purified by boronate affinity.

Ordering information: Quantity: 1µg representing 1.8 nanomoles of 2-300 units of ADP-ribose.

Specifications

Specifications

  • Pk:
    1 µG
  • Cat. No.:
    ENZOALX202043C001
  • Protein/peptide name:
    Poly(ADP-ribose), standard
  • Formulation:
    Liquid. In 13,6 μl water.
  • Purity:
    >95% (HPLC of ADP-ribose RNA and DNA measurement)
  • Concentration:
    131,5 µM

Specifications