Human recombinant PARP1 (catalytic domain) (from Insect cells (Sf9)), HIS Tag
Catalog # ENZOALX201232C020
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:PARP1 (catalytic domain)
- Purity:>90% (SDS-PAGE)
- Concentration:1,5 mg/ml
- Formulation:Liquid. In 50 mM TRIS-HCl, pH 7,5, containing 300 mM sodium chloride, 250 mM imidazole, 2 mM β-mercaptoethanol, 0,2% NP-40 and 10% glycerol.
Specifications
About this item
Produced in SF21 cells. Human PARP-1 (aa 656-1014) is fused to a His-tag.