Order Entry
Slovakia
ContactUsLinkComponent
Recombinant GroES (from E. coli)
Recombinant GroES (from E. coli)
Catalog # ENZOALX201142C025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Recombinant GroES (from E. coli)
Catalog # ENZOALX201142C025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Seq:
    02
  • Protein/peptide name:
    GroES
  • Purity:
    >99%
  • Concentration:
    4,2 mg/ml
  • Formulation:
    Liquid. In 50 mM TRIS/HCl, pH 8,0, containing 10 mM KCl and 10 mM MgCl₂.

Specifications

About this item

Produced in E. coli.