Human recombinant MMP2 proenzyme (from insect cells (Sf9))
Catalog # ENZOALX201095C010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Seq:02
- Protein/peptide name:MMP2 proenzyme
- Purity:>95% (SDS-PAGE)
- Concentration:0,2 mg/ml
- Formulation:Liquid. In 50 mM TRIS-HCl, pH 7,0, 200 mM NaCl, 5 mM CaCl₂, 0,05% Brij-35.
Specifications
About this item
Produced in Sf9 cells.
Active MMP-2 is used to study the degradations of proteins of the extracellular matrix, including fibrillar collagens. MMP-2 is inhibited by tissue inhibitors of matrix metalloproteinases (TIMP), alpha2-macroglobulin and by chelators of divalent cations as EDTA or o-phenanthroline.