Hamster recombinant Grp78 (from E. coli)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Produced in E. coli.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Hamster
- Protein synonyms:BiP
- Protein/peptide name:Grp78
- Purity:-
- Formulation:-
- Purification:-
Specifications
Frequently Bought Together