Bovine native alpha B-Crystallin (from Eye lens)
ENZOADISPP227F
:
- Pk:200 µG
- Protein/peptide type:Native
- Source:Eye lens
- Species:Bovine
- Seq:03
- Protein/peptide name:alpha B-Crystallin
Isolated from bovine eye lens.
Isolated from bovine eye lens.