Human recombinant Cpn10 (from E. coli)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Produced in E. coli.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Protein/peptide name:Cpn10
- Purity:>90% (SDS-PAGE; Western blot)
- Formulation:Liquid, in 20 mM TRIS, pH 8,0, containing 150 mM sodium chloride
- Purification:Purified by multi-step chromatography
Specifications
Frequently Bought Together