Anti-14-3-3 Sheep Polyclonal Antibody
Catalog # ENZOBMLSA4790025
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:14-3-3 protein
- Antigen symbol:14-3-3
- Clonality:Polyclonal
- Host:Sheep
- Reactivity:Human
- Western blot:Yes
- Seq:03
- Antigen synonyms:beta isoform|14-3-3 alpha|Protein 1054|Alpha polypeptide|Protein kinase C inhibitor protein 1|Tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein|protein kinase C inhibitor protein-1|14-3-3 Protein beta/alpha|brain Protein 14-3-3
- Purification:Affinity purified phospho-specific antibody
- Pk:25 µl
Specifications
About this item
Host: Sheep
- For Western Blot
Recognizes human, mouse, rat, cow and sheep 14-3-3beta and 14-3-3zeta phosphorylated at Ser185.
Type: Primary
Antigen: 14-3-3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity: Human