Penicillin G sodium salt 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69-57-8
:
Formula:C₁₆H₁₇N₂NaO₄S
MDL Number:MFCD00069666
:356.38 g/mol
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- pH:5.0 to 7.5
- Specific optical rotation:+293° to +303° (20°C, 589 nm) (c=2, H2O)
Specification Test Results
Frequently Bought Together