

Manganese(II) sulphate monohydrate Ph. Eur., USP
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10034-96-5
:
Formula:MnSO₄·1H₂O
MDL Number:MFCD00149159
Merck Index:13,05763
Melting Pt:700 °C
Density:2.95 g/cm³ (25 °C)
EINECS:232-089-9
UN:3077
ADR:9,III
:169.02 g/mol
:GHS08
:GHS09
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Conforms (see CoA/CoS)
- Conforms to Ph.Eur./USP:Passes test
- Residual solvents:Passes test
- As (Arsenic):Max. 2 ppm
- Cd (Cadmium):Max. 2 ppm
- Co (Cobalt) :Max. 1 ppm
- Hg (Mercury) :Max. 2 ppm
- Ni (Nickel) :Max. 6 ppm
- Pb (Lead) :Max. 5 ppm
- V (Vanadium) :Max. 10 ppm
- Ph.Eur. specifications: :Max. 50 ppm
- Assay (calculated on ignited) [Ph.Eur.]:99.0 --> 101.0 %
- Identification A:Passes test Ph.Eur.
- Identification B:Passes test Ph.Eur.
- Identification C:Passes test Ph.Eur.
- Appearance of solution:Passes test Ph.Eur.
- Cl (Chloride) [Ph.Eur.]:Max. 100 ppm
- Fe (Iron) [Ph.Eur.]:Max. 10 ppm
- Zn (Zinc) [Ph.Eur.]:Max. 50 ppm
- Loss on ignition (500 ± 50°C) [Ph.Eur.]:10.0 --> 12.0 %
- Assay [USP]:98.0 --> 102.0 %
- Not precipitated by (NH4)2S [USP]:Max. 0.5 %
- Loss on ignition (450°C) [USP]:10.0 --> 13.0 %
Specification Test Results
Related Information
Certificates
Search by Lot Number6512_MANGANESE (II) SULPHATE 1H2O PH.EUR. US_BSE_REV0
BSE/TSE Certificates
Manganese(II) Sulphate Monohydrate Ph. Eur., USP
Additional Product Details
sds_VWRC6512
SDS
spec_VWRC6512.1000 spec_VWRC6512.1000 spec_VWRC6512.1000 spec_VWRC6512.9012 spec_VWRC6512.9012
Product specification
More
VWR Chemicals for Biopharma and Pharma Production
Product Literature
Frequently Bought Together