Organic reference standards, Multi-nutrient fertiliser - elements
Catalog # PROONIST-695
Supplier: PROMOCHEM
CAS Number:
Specifications
- Pk:70 g
- Standards:Reference standard
- Description:Multi-nutrient fertiliser - elements
Specifications
About this item
These standards are intended primarily for use in the evaluation of techniques employed in the analysis of multi-nutrient fertiliser materials and materials of a similar matrix.
One unit consists of approximately 70 g of jet-milled fertiliser.
Certifications: Traceable to NIST and SRM.