Human recombinant Allograft Inflammatory Factor 1 (from E. coli)
Catalog # PRSI92-087
Supplier: PROSCI
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:AIF-1|Ionized Calcium-Binding Adapter Molecule 1|Protein G1|Allograft Inflammatory Factor 1|AIF1|IBA1|G1
- Protein/peptide name:Allograft Inflammatory Factor 1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:17.7 kD
- Sequence:Ser2-Pro147
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Allograft Inflammatory Factor 1 (AIF1) contains two EF-hand domains and exists as a homodimer. AIF1 can be detected in T-lymphocytes and peripheral blood mononuclear cells. AIF1 functions as actin-binding protein that enhances membrane ruffling and RAC activation and can enhance the actin-bundling activity of LCP1. In addition, AIF1 plays a role in RAC signaling and in phagocytosis and may also in macrophage activation and function. AIF1 promotes the proliferation of vascular smooth muscle cells and of T-lymphocytes and plays a role in vascular inflammation.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag