Human recombinant STAT3 (from E. coli)
Catalog # PRSI92-085
Supplier: PROSCI
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Signal Transducer and Activator of Transcription 3|Acute-Phase Response Factor|STAT3|APRF
- Protein/peptide name:STAT3
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:21.8 kD
- Sequence:Met1-Asn175
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Signal Transducer and Activator of Transcription 3 (STAT3) belongs to the transcription factor STAT family. STAT3 contains one SH2 domain and is a transcription factor expressed in most cell types. STAT3 is activated by multiple cytokines and growth factors including: IFN-a, IL-10, IL-6, IL-11, IL-12, IL-2, EGF etc. STAT3 functions as signal transducer and transcription activator that mediates cellular responses to interleukins, KITLG/SCF and other growth factors. In addition, STAT3 may also mediate cellular responses to activated FGFR1, FGFR2, FGFR3 and FGFR4.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag