
(-)-Epicatechin ≥95%, crystalline powder
Catalog # J61218.FX
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:20 mg
Specifications
Specification Test Results
- Colour:White to pale yellow
- Form:Crystalline powder
- Solubility:Soluble in water or alcohol
Specification Test Results
About this item
:
:290.271 g/mol
Melting Pt:240 °C (dec.)
EINECS:207-710-1
:GHS07
Storage Temperature:Refrigerator
CAS Number:490-46-0
Formula:C₁₅H₁₄O₆
MDL Number:MFCD00075648