Order Entry
Slovakia
ContactUsLinkComponent
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Flammable
Irritant
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (V/V):
    min. 68.4%
  • Denatonium benzoate (ppm):
    7.9 - 9.2 ppm
  • Methyl ethyl ketone (V/V):
    0.68 - 0.80 %
  • 2-Propanol (V/V):
    0.68 - 0.80%

Specification Test Results

Frequently Bought Together