RNA Purification kits, RNAID® kit, MP Biomedicals
Catalog # ICNA111007200
Supplier: MP BIOMEDICALS
CAS Number:
Specifications
- Pk:1 KIT
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:RNA gel bands
- Sample type:Gel matrix
- Target molecule:RNA
- Description:RNaid® RNA Kit
- Description:RNaid® RNA Kit
Specifications
About this item
The RNaid® Kit is designed to isolate and purify RNA (75 bases to 10 kb) from native and denaturing agarose and polyacrylamide gels.
Ideal for removal of unincorporated nucleotides, buffer salts, and other reagents from enzyme reactions. The purified RNA is ready for reverse transcription and PCR, labeling and microarray analysis, and other common applications.