Order Entry
Slovakia
ContactUsLinkComponent
 
Anti-TSPAN15 Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-9425R-CY7
 :  BIOSS INC
Anti-TSPAN15 Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-9425R-CY7
 :  BIOSS INC

 

  • Antibody type:
    Primary
  • Antigen name:
    Tetraspanin 15
  • Antigen symbol:
    TSPAN15
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    23555
  • Antigen synonyms:
    Tetraspanin 15|Tetraspanin-15|TSPAN15.|TSN15_HUMAN|Transmembrane 4 superfamily member 15|Tetraspan NET-7|Tspan-15
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

NET-7, also known as TSPAN15 (tetraspanin 15) or TM4SF15 (transmembrane 4 superfamily member 15), is a 294 amino acid multi-pass membrane protein that belongs to the transmembrane 4 superfamily, also known as the tetraspanin family. Members of the tetraspanin family are cell-surface proteins that are characterized by the presence of four hydrophobic domains and mediate signal transduction events that play a role in the regulation of cell development, activation, growth, motility, differentiation, and cancer. Considered molecular facilitators, tetraspanin proteins may regulate vesicle fusion and fission.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSPAN15
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat