- Antibody type:Primary
- Antigen name:hydroxyacylglutathione hydrolase-like
- Antigen symbol:HAGHL
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:84264
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
The hydroxyacylglutathione hydrolase-like protein (HAGHL) is a 290 amino acid protein that belongs to the glyoxalase II family. HAGHL binds two zinc ions per subunit and acts as a hydrolase on ester bonds. The gene encoding HAGHL maps to human chromosome 16, which encodes over 900 genes in approximately 90 million base pairs, makes up nearly 3% of human cellular DNA and is associated with a variety of genetic disorders. The rare disorder Rubinstein-Taybi syndrome is associated with chromosome 16, as is Crohn's disease, a gastrointestinal inflammatory condition that may involve the NOD2 gene. An association with systemic lupus erythematosis and a number of other autoimmune disorders with the pericentromeric region of chromosome 16 has led to the identification of SLC5A11 as a potential autoimmune modifier.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HAGHL
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat