- Antibody type:Primary
- Antigen name:G protein-coupled receptor kinase interacting ArfGAP 1
- Antigen symbol:GIT1
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:28964
- Antigen synonyms:GIT1|ARF GTPase-activating protein GIT1|CAT1|G protein-coupled receptor kinase-interactor 1|GRK-interacting protein 1|CAT-1|ARF GAP GIT1|Cool-associated and tyrosine-phosphorylated protein 1
- Modification:phospho-Tyr545
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Heterotrimeric G protein-mediated signal transduction is a dynamically regulated process with the intensity of signal decreasing over time despite the continued presence of the agonist (1,2). G protein-coupled receptor kinases (GRKs) are activated by activated G protein-coupled receptors, and they function to phosphorylate and inactivate cell surface receptors in the heterotrimeric G protein signaling cascade (3,4). GIT1 (for GRK-interactor 1) and GIT2 are GTPase-activating proteins (GAP) for members of the ADP ribosylation factor (ARF) family of small GTP-binding proteins, which are involved in vesicular trafficking (5,6). GIT1 overexpression results in reduced internalization and resensitization of b2-adrenergic receptor, thus reducing b2-adrenergic receptor signaling (5).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GIT1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat